Learn More
Sino Biological Anti-ERK2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50445T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ERK2 / MAPK1 / MAPK2 (Catalog#50445-M20B; P63085; Met1-Ser358). ERK2 / MAPK1 / MAPK2 specific IgG was purified by Mouse ERK2 / MAPK1 / MAPK2 affinity chromatography.
Catalog No. 50-240-8259