Learn More
American Research Products Inc Mouse MatriX Gla Protein MGP
Catalog No. 5000326805
Encompass
Mouse MatriX Gla Protein (MGP) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26805 Supplier American Research Products Inc Supplier No. CSBE16540M
Mouse MatriX Gla Protein (MGP) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich