Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 15 (MMP15) ELISA Kit
Supplier:  Biomatik Corporation EKU05844
Encompass_Preferred
Mouse Matrix Metalloproteinase 15 (MMP15) ELISA Kit for detecting MMP15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54732. Gene ID: 17388. Target Synonyms: SMCP-2; MTMMP2; MT2-MMP; Membrane-Inserted; Membrane-type matrix metalloproteinase 2; Membrane-type-2 matrix metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7305