Learn More
BIOHIPPO Mouse MBP Microsample
SDP
Catalog No. 5000314238
Encompass
Mouse MBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MBP in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 107 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14238 Supplier BIOHIPPO Supplier No. BHE15211764
Mouse MBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MBP in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 107 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color