Learn More
Raybiotech Inc Mouse MCAM/CD146 ELISA Kit
Supplier:  Raybiotech Inc ELMMCAM1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.18 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse MCAM.
Catalog No. 50-195-0078