Learn More
ACROBiosystems ACROBiosystems Mouse MCAM / CD146 Protein, His Tag
Catalog No. 502106510
Encompass_Preferred
Recombinant Protein;100UG;Mouse MCAM, His Tag (MCM-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Val 563 (Accession # Q8R2Y2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.7 kDa. The protein migrates as 70-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6510 Supplier ACROBiosystems Supplier No. MCMM52H3100UG
Recombinant Protein;100UG;Mouse MCAM, His Tag (MCM-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Val 563 (Accession # Q8R2Y2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.7 kDa. The protein migrates as 70-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.