Learn More
Sino Biological Recombinant Anti-CD146/MCAM Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50794R241A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD146 / MCAM (rM CD146 / MCAM; Catalog#50794-M08H; Q8R2Y2-1; Met1-Val563) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-3090