Learn More
Sino Biological Recombinant Anti-CD146/MCAM Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 50794R241F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD146/MCAM (rM CD146/MCAM; Catalog#50794-M08H; Q8R2Y2-1; Met1-Val563) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-3087