Learn More
Raybiotech Inc Mouse MCP-1 ELISA Kit
Supplier:  Raybiotech Inc ELMMCP11
Encompass_Preferred
RayBio Mouse MCP-1 (CCL2) ELISA Kit for cell culture supernatants, plasma, and serum samples.
Catalog No. NC1977992