Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse MCPT-1 (mMCP-1) Uncoated ELISA Kit with Plates

Catalog No. 5017417
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-174-17 10 x 96 Tests
50-174-16 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-174-17 Supplier Invitrogen™ Supplier No. 88750386
  • $876.00
    $832.65 / Each of 1

    Save $43.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Mouse MCPT-1 (mMCP-1) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse MCPT-1. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

MCPT-1 is the only chymase expressed by intestinal mucosal mast cells, which are found in the intestinal epithelium. Although it is expressed constitutively and is detectable in the sera of normal mice, parasites in the gut cause systemic levels to increase dramatically within two days and peak at two weeks following infection. It plays an important role in host defense against intestinal parasites, as mice deficient in MCPT-1 display delayed response upon infection. Elevated MCPT-1 levels are also observed during intestinal allergic hypersensitivity reactions. The mechanism of its action is not fully understood, although it is believed to increase intestinal permeability similarly to rMCPT-II, possibly by cleaving the proteins at tight junctions between cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11034
Assay Range 234-15000 pg/mL
Assay Sensitivity 234 pg/mL
Content And Storage Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant protein for generating standard curve and calibrating samples
Coating Buffer: 10X PBS ELISA Coating Buffer
Assay Diluent: 5X Concentrated
Detection Enzyme: Pre-titrated Avidin-HRP
Substrate Solution: Tetramethylbenzidine (TMB) Solution
Certificate of Analysis: Lot-specific instructions for the dilution of antibodies and standards
96-Well Plates: Corning Costar flat-bottom plated (included with catalog numbers ending in -22 and -86)
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit with Plates
Product Type ELISA
Sample Type Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AV080368; mast cell protease 1; Mcp-1; Mcpt1; mMCP-1AV080368; mast cell protease 1; Mcp-1; Mcpt1; mMCP-1; CSIF; GVHDS; IL-10A; TGIF; IL10
Gene ID (Entrez) 17224
Gene Symbol Mcpt1
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye Biotin
Protein Aliases mMCP-1
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 100 μL, Supernatant 100 μL
Storage Requirements 2-8°C
Synonym Mouse Mast Cell Protease-1
Target MCPT-1
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.