Learn More
Sino Biological Anti-Mast Cell Protease-1/MCPT-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50220RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse MCPT1 (rM MCPT1; Catalog#50220-M08H; NP_032596.1; Met 1-Lys 246). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3366