Learn More
BIOHIPPO Mouse MCT Microsample
SDP
Catalog No. 5000314319
Encompass
Mouse MCT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MCT in serum plasma tissue homogenates It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MCT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MCT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MCT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14319 Supplier BIOHIPPO Supplier No. BHE15212356
Mouse MCT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MCT in serum plasma tissue homogenates It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MCT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MCT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MCT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color