Learn More
Biomatik Corporation Mouse Melanoma Antigen Family B16 (MAGEB16) ELISA Kit
Supplier:  Biomatik Corporation EKU11064
Encompass_Preferred
Mouse Melanoma Antigen Family B16 (MAGEB16) ELISA Kit for detecting MAGEB16 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CWV4. Gene ID: 71967. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4419