Learn More
Biomatik Corporation Mouse Melanoma Associated ME20 (ME20M) ELISA Kit
Supplier:  Biomatik Corporation EKU11065
Encompass_Preferred
Mouse Melanoma Associated ME20 (ME20M) ELISA Kit for detecting ME20M in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60696. Gene ID: 20431. Target Synonyms: PMEL; GP100; SI; SIL; SILV; Pmel17; Silver Homolog; Premelanosome Protein; Silver Locus Protein Homolog; 95 kDa melanocyte-specific secreted glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8234