Learn More
Biomatik Corporation Mouse Melanoma Inhibitory Activity Protein 1 (MIA1) ELISA Kit
Supplier:  Biomatik Corporation EKU05897
Encompass_Preferred
Mouse Melanoma Inhibitory Activity Protein 1 (MIA1) ELISA Kit for detecting MIA1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 37pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61865. Gene ID: 12587. Target Synonyms: CD-RAP; Melanoma-derived growth regulatory protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6704