Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Melatonin R1A/MT1/MTNR1A (NP_032665) VersaClone cDNA

Catalog No. RDC2968
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2968 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2968 Supplier R&D Systems™ Supplier No. RDC2968
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

Specifications

Gene Melatonin R1A/MT1/MTNR1A
Accession Number NP_032665
Gene ID (Entrez) 4543
Gene Alias Mel1a receptor, MEL-1 A-R, melatonin receptor 1 A, melatonin receptor type 1 A, MelatoninR1A, MT1, MTNR1A
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.