Learn More
R&D Systems™ Mouse Melatonin R1B/MT2/MTNR1B (NP_663758.2) VersaClone cDNA

Specifications
Specifications
| Gene | Melatonin R1B/MT2/MTNR1B |
| Accession Number | NP_663758.2 |
| Gene ID (Entrez) | 34632 |
| Gene Alias | CG10692, dDNMT, dDnmt2, Dmel\CG10692, DmMT2, Dnmt, dnmt2, Mel-1 B-R, MelatoninR1B, MT2, Mt2 Methyltransferase 2, MTNR1B, nmt2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.