Learn More
Sino Biological Anti-Melanotransferrin/MFI2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50994T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse MFI2 (Catalog#50994-M08H; Q9R0R1; Met1-Gln708). MFI2 specific IgG was purified by Mouse MFI2 affinity chromatography.
Catalog No. 50-240-8126