Learn More
ACROBiosystems ACROBiosystems Mouse MERTK / Mer Protein, Fc Tag
Catalog No. 502106407
Encompass_Preferred
Recombinant Protein;100UG;Mouse MERTK, Fc Tag (MEK-M5253) is expressed from human 293 cells (HEK293). It contains AA Gly 19 - Met 497 (Accession # Q60805-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 78.7 Kda.
Catalog No. 50-210-6407 Supplier ACROBiosystems Supplier No. MEKM5253100UG
Recombinant Protein;100UG;Mouse MERTK, Fc Tag (MEK-M5253) is expressed from human 293 cells (HEK293). It contains AA Gly 19 - Met 497 (Accession # Q60805-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 78.7 Kda.