Learn More
ACROBiosystems Mouse Mesothelin 100ug
Catalog No. 502534411
Encompass_Preferred
Mouse Mesothelin Protein Fc Tag (MSN-M5254) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).
Catalog No. 50-253-4411 Supplier ACROBiosystems Supplier No. MSNM5254100UG
Mouse Mesothelin Protein Fc Tag (MSN-M5254) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).