Learn More
Meso Scale Discovery Mouse Metabolic Kit 20 Plate
SDP
Catalog No. 5000080607
Encompass
The Mouse Metabolic Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring metabolic biomarkers in mouse samples The kit contains a 96-well plate coated with capture antibodies for insulin leptin resistin PAI-1 total and TNF-alpha It also includes detection antibodies calibrators controls buffers and wash solutions needed to run the assay The kit allows simultaneous quantitation of the 5 metabolic biomarkers from a single small sample volume It is for research use only
Catalog No. 50-000-80607 Supplier Meso Scale Discovery Supplier No. K15124C3
Add to Cart
Add to Cart
Request a Quote
The Mouse Metabolic Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring metabolic biomarkers in mouse samples The kit contains a 96-well plate coated with capture antibodies for insulin leptin resistin PAI-1 total and TNF-alpha It also includes detection antibodies calibrators controls buffers and wash solutions needed to run the assay The kit allows simultaneous quantitation of the 5 metabolic biomarkers from a single small sample volume It is for research use only