Learn More
American Research Products Inc Mouse Meteorin like protein M
Catalog No. 5000327110
Encompass
Mouse Meteorin-like protein (METRNL) ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27110 Supplier American Research Products Inc Supplier No. CSBEL013718MO
Mouse Meteorin-like protein (METRNL) ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich