Learn More
Biomatik Corporation Mouse Methionine Adenosyltransferase I Alpha (MAT1a) CLIA Kit
Supplier:  Biomatik Corporation EKU10020
Encompass_Preferred
Mouse Methionine Adenosyltransferase I Alpha (MAT1a) CLIA Kit for detecting MAT1a in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 68.6-50,000pg/ml. Sensitivity: 29.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q91X83. Gene ID: 11720. Target Synonyms: MAT; SAMS; MATA1; SAMS1; AMS1; S-Adenosylmethionine Synthetase; S-adenosylmethionine synthase isoform type-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4677