Learn More
Biomatik Corporation Mouse Methylthioadenosine Phosphorylase (MTAP) ELISA Kit
Supplier:  Biomatik Corporation EKU05941
Encompass_Preferred
Mouse Methylthioadenosine Phosphorylase (MTAP) ELISA Kit for detecting MTAP in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CQ65. Gene ID: 66902. Target Synonyms: MSAP; S-methyl-5'-thioadenosine phosphorylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8872