Learn More
BIOHIPPO Mouse MHCDRb1 Microsample
SDP
Catalog No. 5000315173
Encompass
Mouse MHCDRb1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MHCDRb1 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 39 07-2500 pg/mL with a sensitivity of 12 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MHCDRb1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MHCDRb1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MHCDRb1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15173 Supplier BIOHIPPO Supplier No. BHE15210872
Mouse MHCDRb1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MHCDRb1 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 39 07-2500 pg/mL with a sensitivity of 12 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MHCDRb1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MHCDRb1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MHCDRb1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color