Learn More
Litron Laboratories MicroFlow PLUS-M Kit
Catalog No. NC0844665
Encompass
Analyze mouse blood samples for micronuclei using your own flow cytometer. Each MicroFlow PLUS-M kit contains all of the quality-assured supplies & reagents to collect and fix 60 mouse blood samples. With this MicroFlow PLUS kit, analyses are performed using your own in-house flow cytometer.
Catalog No. NC0844665 Supplier Litron Laboratories Supplier No. PLUSM
Analyze mouse blood samples for micronuclei using your own flow cytometer. Each MicroFlow PLUS-M kit contains all of the quality-assured supplies & reagents to collect and fix 60 mouse blood samples. With this MicroFlow PLUS kit, analyses are performed using your own in-house flow cytometer.