Learn More
Sciencell Research Laboratories Mouse Microglia, C57BL/6, Cryopreserved P0, >1 x 10^6 cells, 1 ml vial, Immunofluorescence: F4/80, CD11b
Catalog No. NC0757591
Encompass
ScienCell Research Laboratories' Mouse Microglia are primary cells sourced from the brains of postnatal day 2 C57BL/6 mice. These cryopreserved cells (passage 0) can be directly utilized in experiments post-thawing, providing researchers with a valuable resource for exploring neuroinflammation and brain pathology. These cells are rigorously tested for contaminants, ensuring quality and reliable experimental outcomes.
  • Derived from C57BL/6 mouse brain, ensuring high physiological relevance for in vitro studies.
  • Guaranteed negative for mycoplasma, bacteria, yeast, and fungi.
  • Cryopreserved at passage 0 for immediate experimental use after thawing.
  • Well-characterized source isolated from postnatal day 2 mice.
  • Immunophenotypes confirmed with F4/80 and CD11b markers.
  • Quality controlled for research-intensive applications.
Catalog No. NC0757591 Supplier Sciencell Research Laboratories Supplier No. M190057
ScienCell Research Laboratories' Mouse Microglia are primary cells sourced from the brains of postnatal day 2 C57BL/6 mice. These cryopreserved cells (passage 0) can be directly utilized in experiments post-thawing, providing researchers with a valuable resource for exploring neuroinflammation and brain pathology. These cells are rigorously tested for contaminants, ensuring quality and reliable experimental outcomes.
  • Derived from C57BL/6 mouse brain, ensuring high physiological relevance for in vitro studies.
  • Guaranteed negative for mycoplasma, bacteria, yeast, and fungi.
  • Cryopreserved at passage 0 for immediate experimental use after thawing.
  • Well-characterized source isolated from postnatal day 2 mice.
  • Immunophenotypes confirmed with F4/80 and CD11b markers.
  • Quality controlled for research-intensive applications.