Learn More
Biomatik Corporation Mouse Midline 1 (MID1) ELISA Kit
Supplier:  Biomatik Corporation EKU05962
Encompass_Preferred
Mouse Midline 1 (MID1) ELISA Kit for detecting MID1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70583. Gene ID: 17318. Target Synonyms: BBBG1; Midin; FXY; GBBB1; OGS1; OS; OSX; RNF59; TRIM18; XPRF; ZNFXY; Opitz/BBB Syndrome; RING finger protein 59; Tripartite motif-containing protein 18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8869