Learn More
Sino Biological Anti-MIF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50066T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse MIF (Catalog#50066-M08H; P34884; Pro2-Ala115). MIF specific IgG was purified by Mouse MIF affinity chromatography.
Catalog No. 50-255-3497