Learn More
Sino Biological Recombinant Mouse Allergin 1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 54920M02H100
Encompass_Preferred
A DNA sequence encoding the mouse MILR1 (NP_001028607.1) (Met1-Ser150) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2960