Learn More
Cayman Chemical Mouse MInralocortIcoId Recep
SDP
Catalog No. 502942891
Encompass_Preferred
INDIGO Bioscience’s Assays are all-inclusive cell-based assay systems. In addition to mouse MR Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist Aldosterone, Luciferase Detection Reagent, and a cell culture-ready assay plate.
Catalog No. 50-294-2891 Supplier Cayman Chemical Supplier No. 4104996 WELL
INDIGO Bioscience’s Assays are all-inclusive cell-based assay systems. In addition to mouse MR Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist Aldosterone, Luciferase Detection Reagent, and a cell culture-ready assay plate.