Learn More
Raybiotech Inc Mouse MIP-1 beta/CCL4 ELISA Kit
Supplier:  Raybiotech Inc ELMMIP1B1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse MIP-1 beta.
Catalog No. 50-195-0089