Learn More
FUJIFILM BIOSCIENCES INC Mouse MIP-1 beta (CCL4), Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Catalog No. 50900526
Encompass_Preferred
Macrophage inflammatory protein-1 beta (MIP-1 B), also known as CCL4, is  produced by macrophages and functions as a mitogen-inducible cytokine.  MIP-1 B signals through the chemokine receptor CCR5 to chemoattract immune cells.  MIP-1 B induces inflammatory responses, including neutrophil superoxide production.  The MIP-1 B and MIP-1 A heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1). 
Catalog No. 50-900-526 Supplier FUJIFILM BIOSCIENCES INC Supplier No. 20043100UG
Add to Cart
Add to Cart
Macrophage inflammatory protein-1 beta (MIP-1 B), also known as CCL4, is  produced by macrophages and functions as a mitogen-inducible cytokine.  MIP-1 B signals through the chemokine receptor CCR5 to chemoattract immune cells.  MIP-1 B induces inflammatory responses, including neutrophil superoxide production.  The MIP-1 B and MIP-1 A heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1). 