Learn More
FUJIFILM Biosciences Mouse MIP-3 beta (CCL19), Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Catalog No. 50990875
Encompass_Preferred
Macrophage inflammatory protein-3 beta (MIP-3 B), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes, and activated bone marrow stromal cells.  MIP-3 B signals through the G protein-coupled receptor CCR7 to regulate normal lymphocyte recirculation.  MIP-3 B also functions  during T cell trafficking to the thymus, and in T cell and B cell homing to the lymph nodes and secondary lymphoid organs.  
Catalog No. 50-990-875 Supplier FUJIFILM Biosciences Supplier No. 20044100UG
Macrophage inflammatory protein-3 beta (MIP-3 B), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes, and activated bone marrow stromal cells.  MIP-3 B signals through the G protein-coupled receptor CCR7 to regulate normal lymphocyte recirculation.  MIP-3 B also functions  during T cell trafficking to the thymus, and in T cell and B cell homing to the lymph nodes and secondary lymphoid organs.  