Learn More
FUJIFILM BIOSCIENCES INC Mouse MIP-3 alpha (CCL20), Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 20051100UG
Encompass_Preferred
Macrophage inflammatory protein-3 alpha (MIP-3 A), also called CCL20, is expressed in the liver, lungs, lymph nodes, and peripheral blood lymphocytes.  MIP-3 A expression is strongly induced by inflammatory signals, and downregulated by the anti-inflammatory cytokine interleukin 10 (IL-10).  MIP-3 A signals through the G protein-coupled receptor CCR6 to function as a chemoattractant to lymphocytes and dendritic cells. 
Catalog No. 50-990-894