Learn More
BIOHIPPO Mouse MIP3b Microsample
SDP
Catalog No. 5000314523
Encompass
Mouse MIP3b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MIP3b in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MIP3b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MIP3b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MIP3b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14523 Supplier BIOHIPPO Supplier No. BHE15212094
Mouse MIP3b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MIP3b in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MIP3b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MIP3b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MIP3b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color