Learn More
Biomatik Corporation Mouse Misato Homolog 1 (MSTO1) ELISA Kit
Supplier:  Biomatik Corporation EKU05970
Encompass_Preferred
Mouse Misato Homolog 1 (MSTO1) ELISA Kit for detecting MSTO1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2YDW2. Gene ID: 229524. Target Synonyms: MST. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8867