Learn More
American Research Products Inc Mouse Mitochondrial brown fat
Catalog No. 5000327685
Encompass
Mouse Mitochondrial brown fat uncoupling protein 1 (UCP1) ELISA kit from CUSABIO Detection Range 4 68 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27685 Supplier American Research Products Inc Supplier No. CSBEL025554MO
Mouse Mitochondrial brown fat uncoupling protein 1 (UCP1) ELISA kit from CUSABIO Detection Range 4 68 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich