Learn More
Biomatik Corporation Mouse Mitochondrial Methionyl tRNA Formyltransferase (MTFMT) ELISA Kit
Supplier:  Biomatik Corporation EKU05971
Encompass_Preferred
Mouse Mitochondrial Methionyl tRNA Formyltransferase (MTFMT) ELISA Kit for detecting MTFMT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D799. Gene ID: 69606. Target Synonyms: FMT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7520