Learn More
ACROBiosystems Biotinylated Monoclonal Anti-MMAE&MMAF Antibody, Mouse IgG1 (MALS verified)
Catalog No. 502719144
Encompass_Preferred
Monomethyl auristatin E (MMAE) is a synthetic derivative of dolastatin 10 and functions as a potent mitotic inhibitor by inhibiting tubulin polymerization. MMAE is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types. Anti-MMAE antibody is a Mouse monoclonal antibody. This antibody has been shown to work in applications such as: PK, PD, Immunoassay and ELISA.
Catalog No. 50-271-9144 Supplier ACROBiosystems Supplier No. MMEBLS10425UG
Monomethyl auristatin E (MMAE) is a synthetic derivative of dolastatin 10 and functions as a potent mitotic inhibitor by inhibiting tubulin polymerization. MMAE is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types. Anti-MMAE antibody is a Mouse monoclonal antibody. This antibody has been shown to work in applications such as: PK, PD, Immunoassay and ELISA.