Learn More
ACROBiosystems HRP conjugated Anti-MMAE Antibody, mAb
Catalog No. 502331457
Encompass_Preferred
HRP conjugated Anti-MMAE Antibody, is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Bevacizumab MMAE.
Catalog No. 50-233-1457 Supplier ACROBiosystems Supplier No. MMEPLS10425UG
HRP conjugated Anti-MMAE Antibody, is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Bevacizumab MMAE.