Learn More
Raybiotech Inc Mouse MMP-10 ELISA Kit
Supplier:  Raybiotech Inc ELMMMP101
Encompass_Preferred
For Cell Culture Supernatants, Serum. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse MMP-10.
Catalog No. 50-195-0094