Learn More
Sino Biological Mouse MMP8 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50493MNAH50
Encompass_Preferred
A DNA sequence encoding the pro form of mouse MMP8 (NP_032637.3) (Phe 21-Ser 465) was expressed and purified, with an initial Met at the N-terminus.
Catalog No. 50-237-3598