Learn More
Sino Biological Anti-MNX1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106653T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse MNX1, and purified by antigen affinity chromatography.
Catalog No. 50-241-1489