Learn More
ACROBiosystems Mouse MOG 25ug
Supplier:  ACROBiosystems MOGM82E725UG
Encompass_Preferred
Biotinylated Mouse MOG Protein HisAvitag (MOG-M82E7) is expressed from human 293 cells (HEK293). It contains AA Gly 29 -Thr 156 (Accession # Q61885).
Catalog No. 50-253-4416