Learn More
Biolegend MojoSort™ Mouse CD4 T Cell Isolation Kit
Supplier:  Biolegend 480005
Encompass_Preferred
MojoSort™ Mouse CD4 T Cell Isolation Kit Reactivity: Mouse; Apps: Cell Sep - Neg; Size: 10 tests
Catalog No. NC1726424