Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mouse monoclonal [B3102E8] Anti-Human IgE Fc (Biotin), 250 ug

Catalog No. 502653619
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-3619 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-3619 Supplier Abcam Supplier No. AB99807250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse monoclonal [B3102E8] Anti-Human IgE Fc (Biotin)

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: Dot, ELISA, ICC, IHC-Fr, WB
  • Species Reactivity: Human
  • Storage Buffer: PBS
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Applications Dot Blot, Western Blot, Immunocytochemistry, Immunohistochemistry (Frozen), ELISA
Classification Monoclonal
Clone B3102E8
Conjugate Biotin
Formulation pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
Gene Symbols IGHE
Host Species Mouse
Purification Method Affinity purification
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG1
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv Dot, WB, ICC, IHC-Fr, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.