Learn More
Abcam Mouse monoclonal [B3102E8] Anti-Human IgE Fc (HRP), 500 uL
Description
- Host Species: Mouse
- Clonality: Monoclonal
- Isotype: IgG1
- Applications: Dot, ELISA, ICC, IHC-Fr, WB
- Species Reactivity: Human
- Storage Buffer: PBS, Glycerol
- Form: Liquid
- Purification: Proprietary technique
Specifications
Specifications
| Applications | Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | B3102E8 |
| Conjugate | HRP |
| Formulation | pH: 7.4, Constituents: PBS, 50% Glycerol (glycerin, glycerine) |
| Gene Symbols | IGHE |
| Host Species | Mouse |
| Purification Method | Proprietary technique |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.