Learn More
Abcam Mouse monoclonal [JDC-10] Anti-Human IgG Fc (PE), 100 ug
Description
- Host Species: Mouse
- Clonality: Monoclonal
- Isotype: IgG1
- Applications: Dot, ELISA, ICC/IF, IHC-Fr, WB
- Species Reactivity: Human
- Storage Buffer: PBS, Sucrose
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | JDC-10 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.3, Preservative: 0.1% Sodium azide, Constituents: Sucrose, PBS |
| Gene | IGHG1 |
| Gene Symbols | IGHG1 |
| Host Species | Mouse |
| Purification Method | Affinity purification |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.