Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mouse monoclonal [JDC-10] Anti-Human IgG Fc, 250 ug

Catalog No. 502653628
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-3628 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-3628 Supplier Abcam Supplier No. AB99757250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse monoclonal [JDC-10] Anti-Human IgG Fc

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: Dot, ELISA, ICC, ICC/IF, IHC-Fr, WB
  • Species Reactivity: Human
  • Storage Buffer: Borate buffered saline
  • Form: Liquid
  • Purification: Affinity purification Protein G

Specifications

Applications Immunocytochemistry, Immunofluorescence, Dot Blot, ELISA, Western Blot, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone JDC-10
Conjugate Unconjugated
Formulation pH: 8.2, Constituents: 100% Borate buffered saline
Gene Symbols IGHG1
Host Species Mouse
Purification Method Affinity purification Protein G
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG1
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv ICC/IF, Dot, ELISA, WB, ICC, IHC-Fr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.